Skip to Content
Merck
All Photos(1)

Key Documents

SRP2094

Sigma-Aldrich

Fos-related antigen 1 human

recombinant, expressed in insect cells, ≥70% (SDS-PAGE)

Synonym(s):

FRA, FRA1, fra-1

Sign Into View Organizational & Contract Pricing

Select a Size

5 μG
$445.00

$445.00


Estimated to ship on26 May 2025


Request a Bulk Order

Select a Size

Change View
5 μG
$445.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.26

$445.00


Estimated to ship on26 May 2025


Request a Bulk Order

biological source

human

recombinant

expressed in insect cells

Assay

≥70% (SDS-PAGE)

form

frozen liquid

mol wt

~31.2 kDa

packaging

pkg of 5 μg

storage condition

avoid repeated freeze/thaw cycles

concentration

350 μg/mL

color

colorless to clear

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... FOSL1(8061)

Biochem/physiol Actions

The fos-related antigen 1 (Fra-1, or FosL1: Fos-like antigen 1) is a member of the Fos onco-protein family and a component of AP-1 transcription factor. Similar to other members of the family, Fra-1 contains a leucine zipper domain and a highly conserved C-terminal region. Transcription of Fra-1 gene is regulated by multiple cis-elements and 12-O-tetradecanoylphorbol-13-acetate (TPA). Fra-1 expression is associated with proliferation and invasiveness of breast cancer cells and other cancer cells.

Physical form

Clear and colorless frozen liquid solution

Preparation Note

Use a manual defrost freezer and avoid repeated freeze-thaw cycles. While working, please keep sample on ice.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D R Cohen et al.
Genes & development, 3(2), 173-184 (1989-02-01)
fra-1 encodes a serum-inducible protein (Fra-1) that is antigenically related to Fos. We have characterized Fra-1 expression in serum-stimulated cells using antibodies raised against several regions of this protein. Fra-1, expressed transiently in COS cells or in serum-stimulated rat fibroblasts
Pavan Adiseshaiah et al.
The Journal of biological chemistry, 278(48), 47423-47433 (2003-09-19)
Recent studies indicate a potential role for Fra-1, a heterodimeric partner of activator protein 1 (AP1), in toxicant-induced epithelial injury, repair, and cellular transformation. Here, we have investigated the transcriptional regulation of fra-1 by 12-O-tetradecanoylphorbol-13-acetate (TPA) in human bronchial epithelial
M Matsui et al.
Oncogene, 5(3), 249-255 (1990-03-01)
cDNA clones of human fos-related genes fra-1 and fra-2 were isolated by screening human c-DNA libraries with human fos DNA as a probe. We obtained human fra-1 cDNA clones that can code for a protein of 271 amino acid residues

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service