Skip to Content
Merck
All Photos(1)

Key Documents

SRP2006

Sigma-Aldrich

TFIIE, α+β subunits human

recombinant, expressed in E. coli, ≥80% (SDS-PAGE)

Synonym(s):

FE, TF2E1, TF2E2, TFIIE-A, TFIIE-B

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
$587.00

$587.00


Estimated to ship on22 May 2025



Select a Size

Change View
10 μG
$587.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.26

$587.00


Estimated to ship on22 May 2025


biological source

human

recombinant

expressed in E. coli

Assay

≥80% (SDS-PAGE)

form

frozen liquid

mol wt

~111 kDa

packaging

pkg of 10 μg

storage condition

avoid repeated freeze/thaw cycles

concentration

500 μg/mL

color

clear colorless

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

Biochem/physiol Actions

The human Transcription Factor IIE (TFIIE) is composed of 56 kDa (α) and 34 kDa (β) subunits and is shown to be a heterotetramer. The 56 kDa subunit contains a region similar to a zinc-binding domain and a region sharing homology with the catalytic loop of a kinase domain. TFIIE binds to RNA polymerase II in solution and joins the pre-initiation complex probably concomitant with RNA polymerase II and TFIIF.

Physical form

Clear and colorless frozen liquid solution

Preparation Note

Use a manual defrost freezer and avoid repeated freeze-thaw cycles. While working, please keep sample on ice.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Inostroza et al.
The Journal of biological chemistry, 266(14), 9304-9308 (1991-05-15)
Mammalian RNA polymerase II transcription factor IIE (TFIIE) was purified to apparent homogeneity. The activity copurified with polypeptides of 34 and 56 kDa. The 56-kDa subunit was sufficient for low levels of transcription activity in a transcription system reconstituted in
Y Ohkuma et al.
Proceedings of the National Academy of Sciences of the United States of America, 87(23), 9163-9167 (1990-12-01)
Human transcription factor TFIIE, a ubiquitous factor required for transcription initiation by RNA polymerase II, was purified to homogeneity by a combination of conventional and HPLC steps. The purified TFIIE contained equimolar amounts of 57-kDa (TFIIE-alpha) and 34-kDa (TFIIE-beta) polypeptides
M G Peterson et al.
Nature, 354(6352), 369-373 (1991-12-05)
The general transcription factor IIE (TFIIE) is an essential component of the eukaryotic RNA polymerase II initiation complex. We have isolated human complementary DNA clones for both the subunits of TFIIE. Using purified recombinant proteins we find that both subunits

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service