Skip to Content
Merck
All Photos(2)

Key Documents

SRP0726

Sigma-Aldrich

STK21 (CIT), Active

recombinant

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
$889.00

$889.00


Estimated to ship on15 May 2025



Select a Size

Change View
10 μG
$889.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.77

$889.00


Estimated to ship on15 May 2025


recombinant

expressed in baculovirus infected Sf9 cells

Assay

70-95% (SDS-PAGE)

form

frozen liquid

specific activity

4.0-6.0 nmol/min·mg (radiometric)

mol wt

~74 kDa

concentration

0.1 μg/μL

accession no.

NM_007174

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... STK21(11113)

Application

Useful for kinase assay and Western blot.

Biochem/physiol Actions

Recombinant human STK21 (CIT) (1-449) was expressed by Baculovirus in Sf9 insect cells using an N-terminal GST tag. STK21 is a serine/threonine-protein kinase that functions in cell division and also to promote efficient cytokinesis. STK21 is a novel molecular mechanism for a subset of human malformation syndromes of the central nervous system (1). STK21 is also associated with bipolar disorder and risk for schizophrenia. STK21 is a novel partner for the GTP-bound forms of rho and rac (2).

Reconstitution

Formulated in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25 mM DTT, 0.1 mM PMSF, with 25% glycerol.

Storage and Stability

Store in aliquots at -70°C. Avoid freeze/thaw cycles.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service