Skip to Content
Merck
All Photos(2)

Key Documents

SRP0705

Sigma-Aldrich

IFNA4

recombinant

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
$845.00

$845.00


Estimated to ship on01 June 2025



Select a Size

Change View
50 μG
$845.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

$845.00


Estimated to ship on01 June 2025


recombinant

expressed in E. coli

Assay

70-95% (SDS-PAGE)

form

lyophilized powder

mol wt

~19.6 kDa

accession no.

P05014

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... IFNA4(3441)

Biochem/physiol Actions

Recombinant Human IFNA4 was expressed in E. coli cells. All known subtypes of IFN- alpha show the same antiviral antiparasitic, antiproliferative activities. IFN-alpha forms are produced by monocytes/macrophages, lymphoblastoid cells, fibroblasts, and a number of different cell types following induction by viruses, nucleic acids, glucocorticoid hormones, and low-molecular weight substances. All IFN-alpha subtypes possess a common conserved sequence region between amino acid positions 115-151 while the amino-terminal ends are variable. Many IFN-alpha subtypes differ in their sequences at only one or two positions. IFN-alpha and IFN-beta are thought to bind to the same receptor. Signal transduction mechanisms elicited after binding of IFN-alpha to its receptors involves tyrosine phosphorylation (see also: PTK; protein tyrosine kinase) of various non-receptor tyrosine kinases belonging to the Janus kinases.

Reconstitution

Formulated as a lyophilized protein from a 0.2 um filtered solution in Tris and NaCl, pH 8.0. Upon reconstitution, working aliquots can be stored at 2-8°C for one month, or at -20°C for six months, with a carrier protein without detectable loss of activity.

Storage and Stability

The lyophilized protein is stable for at least one year from date of receipt at -70°C. Upon reconstitution, this cytokine can be stored in working aliquots at 2-8°C for one month, or at -20°C for six months, with a carrier protein without detectable loss of activity. Avoid repeated freeze/thaw cycles.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service