Skip to Content
Merck
All Photos(1)

Key Documents

SRP0679

Sigma-Aldrich

LRRKTIDE

≥95% (HPLC)

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$188.00

$188.00


Check Cart for Availability


Select a Size

Change View
1 MG
$188.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.24

$188.00


Check Cart for Availability

Assay

≥95% (HPLC)

form

lyophilized powder

mol wt

1931.29

shipped in

ambient

storage temp.

−20°C

Application

Useful as a substrate for LRRK kinases.

Biochem/physiol Actions

The LRRKtide peptide sequence (RLGRDKYKTLRQIRQ) is derived from human ezrin (amino acids 561-573), moesin (amino acids 539-553) and radixin (amino acids 558-570).

Sequence

RLGRDKYKTLRQIRQ

Reconstitution

Reconstitute in 20mM Tris-HCl, pH 7.5 solution to a final concentration of 1mg/mL.

Storage and Stability

Store diluted product in aliquots at -20°C. Avoid freeze/thaw cycles. Stability: >1yr at -20°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service