Skip to Content
Merck
All Photos(1)

Key Documents

SRP0624

Sigma-Aldrich

FCRN, His-Tag

recombinant, expressed in HEK 293 cells

Synonym(s):

Brambell receptor, neonatal Fc receptor

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$754.00

$754.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
$754.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.26

$754.00


Check Cart for Availability

Request a Bulk Order

recombinant

expressed in HEK 293 cells

form

frozen liquid

mol wt

32 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... FCGRT(2217)

General description

Human complex of FcRn also known as Neonatal Fc receptor, heterodimer of FCGRT (Fc Fragment of IgG Receptor and Transporter), GenBank Accession No. NM_001136019, a.a. 24-297, with a C-terminal His-tag, MW=32 kDa and B2M, also known as beta 2 microglobulin, GenBank Accession No. NM_004048, a.a 1-119, MW=14 kDa. Individually expressed in a HEK293 expression system.

Physical form

Aqueous solution

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. What are the constituents of the buffer this protein is supplied in? Thanks

    1 answer
    1. The buffer used to prepare this product is 8 mM phosphate, pH 7.4, 110 mM NaCl, 2.2 mM KCl, and 20 % glycerol.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service