Skip to Content
Merck
All Photos(1)

Key Documents

SRP0589

Sigma-Aldrich

SIRP-gamma (CD172G), FC Fusion, Biotin Labeled

recombinant, expressed in HEK 293 cells

Synonym(s):

CD172g, Signal regulatory protein-gamma, signal regulatory protein beta 2

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
$1,010.00

$1,010.00


Check Cart for Availability


Select a Size

Change View
50 μG
$1,010.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.26

$1,010.00


Check Cart for Availability

recombinant

expressed in HEK 293 cells

form

frozen liquid

mol wt

67.6 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... SIRPG(55423)

General description

Signal regulatory proteingamma (SIRP-γγ), also known as signal regulatory protein beta 2, and CD172g, GenBank Accession No. NP_018556, a.a. 29-360, fused at the C-terminus to the Fc portion of human IgG1 and C terminal Avitag , MW 67.6 kDa, expressed in a HEK293 expression system and enzymatically biotinylated using Avitag technology. Note: SIRP-γ is heavily glycosylated, resulting in higher molecular weight. Biotinylation was confirmed to be >90%.

Physical form

Aqueous solution

Legal Information

AviTag is a trademark of Avidity, LLC

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service