Skip to Content
Merck
All Photos(1)

Key Documents

SRP0582

Sigma-Aldrich

Human PD-1, FLAG® - AviTag -His-Tag

recombinant, expressed in HEK 293 cells

Synonym(s):

CD279, HPD-L, PD-1, PDCD1, Programmed cell death 1, SLEB2

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
$762.00

$762.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 μG
$762.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.26

$762.00


Check Cart for Availability

Request a Bulk Order

recombinant

expressed in HEK 293 cells

form

frozen liquid

mol wt

20 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... PDCD1(5133)

General description

Human secreted programmed cell death 1 (PD-1), also known as PDCD1, SLEB2, CD279 and HPD-L, GenBank Accession No. NM_005018, a.a. 25-167, with C-terminal FLAG-Avi-His-tags, MW=20 kDa, expressed in a HEK293 cell expression system. This protein runs at a higher M.W. by SDS-PAGE due to glycosylation.

Physical form

Aqueous solution

Legal Information

AviTag is a trademark of Avidity, LLC
FLAG is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. Could you please provide information on the buffer required for the proteins in SRP0569 and SRP0582?

    1 answer
    1. The solution consists of 8 mM phosphate at pH 7.4, 110 mM NaCl, 2.2 mM KCl, and 20% glycerol.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service