Skip to Content
Merck
All Photos(1)

Key Documents

SRP0574

Sigma-Aldrich

LAT, His-Tag

recombinant, expressed in baculovirus infected Sf9 cells

Synonym(s):

linker for activation of T cells family member 1, phospho-tyrosine adapter protein, pp36

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$741.00

$741.00


Check Cart for Availability


Select a Size

Change View
20 μG
$741.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.26

$741.00


Check Cart for Availability

recombinant

expressed in baculovirus infected Sf9 cells

form

frozen liquid

mol wt

24.4 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... LAT(27040)

General description

Human linker for activation of T cells family member 1 (LAT), also known as 36 kDa phospho-tyrosine adapter protein, pp36. GenBank Accession No. NM_001014989* variant 4, a.a. 64-269(end), with N-terminal His-tag, expressed in a baculovirus-infected Sf9 cell expression system. MW = 24.4 kDa. This protein runs at a higher MW by SDSPAGE due to glycosylation.

Physical form

Aqueous solution

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service