Skip to Content
Merck
All Photos(1)

Key Documents

SRP0481

Sigma-Aldrich

TRIM24 (896-1014) human

recombinant, expressed in E. coli, ≥90% (SDS-PAGE)

Synonym(s):

PTC6, RNF82, TF1A, TIF1, tripartite motif containing 24

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$972.00

$972.00


Estimated to ship on30 May 2025



Select a Size

Change View
100 μG
$972.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

$972.00


Estimated to ship on30 May 2025


biological source

human

recombinant

expressed in E. coli

Assay

≥90% (SDS-PAGE)

form

aqueous solution

mol wt

15 kDa

packaging

pkg of 100 μg

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... TRIM24(8805)

General description

Human bromodomain tripartite motif containing 24 (TRIM24), also known as TIF1 (GenBank Accession No. NM_015905) amino-acids 896 - 1014 with N-terminal HIS-tag, MW = 15 kDa, expressed in an E. coli expression system.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tsai, W.W., et al.
Nature, 468, 927-932 (2012)
Misato Kikuchi et al.
Biochimica et biophysica acta, 1793(12), 1828-1836 (2009-11-17)
The androgen receptor (AR) is a ligand-dependent transcription factor that belongs to the family of nuclear receptors, and its activity is regulated by numerous AR coregulators. AR plays an important role in prostate development and cancer. In this study, we

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service