Skip to Content
Merck
All Photos(1)

Key Documents

SRP0476

Sigma-Aldrich

SMARCA2 (1375-1511) human

recombinant, expressed in E. coli, ≥90% (SDS-PAGE)

Synonym(s):

BRM, SNF2, SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a, member 2, SWI2, Sth1p

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$972.00

$972.00


Check Cart for Availability


Select a Size

Change View
100 μG
$972.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

$972.00


Check Cart for Availability

biological source

human

recombinant

expressed in E. coli

Assay

≥90% (SDS-PAGE)

form

aqueous solution

mol wt

16.9 kDa

packaging

pkg of 100 μg

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... SMARCA2(6595)

General description

Human Bromodomain SMARCA2 or BRM (GenBank Accession No. NM_003070) amino-acids 1375 – 1511 with N-terminal HIS-tag, MW = 16.9 kDa, expressed in an E. coli expression system.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Hunter Shain et al.
Proceedings of the National Academy of Sciences of the United States of America, 109(5), E252-E259 (2012-01-12)
Defining the molecular genetic alterations underlying pancreatic cancer may provide unique therapeutic insight for this deadly disease. Toward this goal, we report here an integrative DNA microarray and sequencing-based analysis of pancreatic cancer genomes. Notable among the alterations newly identified
G Liu et al.
Oncogene, 30(29), 3295-3304 (2011-04-12)
SWI/SNF (SWItch/sucrose non-fermentable) complexes are ATP-dependent chromatin remodeling enzymes critically involved in the regulation of multiple functions, including gene expression, differentiation, development, DNA repair, cell adhesion and cell cycle control. BRM, a key SWI/SNF complex subunit, is silenced in 15-20%

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service