Skip to Content
Merck
All Photos(1)

Key Documents

SRP0402

Sigma-Aldrich

Histone H3 peptide (1-21), biotin

≥90% (HPLC)

Sign Into View Organizational & Contract Pricing

Select a Size

80 NMOL
$442.00

$442.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
80 NMOL
$442.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

$442.00


Check Cart for Availability

Request a Bulk Order

Assay

≥90% (HPLC)

form

aqueous solution

packaging

pkg of 80 nmol

concentration

400 μM

shipped in

dry ice

storage temp.

−70°C

General description

Histone H3 Peptide, biotin conjugate. The peptide corresponds to residues 1-21 of histone H3.

Application

200 μL at 400 μM, or 80 nmol Sold as 80 nmol
Study enzyme kinetics, and screen small molecular inhibitors of Histone 3 (K4,K9) methyltransferases for drug discovery and HTS applications.

Physical form

Supplied in a TRIS-buffered solution.

Other Notes

200 μL at 400 μM, or 80 nmol Sold as 80 nmol

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service