Skip to Content
Merck
All Photos(1)

Key Documents

SRP0378

Sigma-Aldrich

PLU-1 murine

recombinant, expressed in baculovirus infected Sf9 cells, ≥50% (SDS-PAGE)

Synonym(s):

JARID1B, KDM5B, lysine (K)-specific demethylase 5B

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$972.00

$972.00


Check Cart for Availability


Select a Size

Change View
20 μG
$972.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

$972.00


Check Cart for Availability

biological source

rat

recombinant

expressed in baculovirus infected Sf9 cells

Assay

≥50% (SDS-PAGE)

form

aqueous solution

mol wt

86 kDa

packaging

pkg of 20 μg

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

mouse ... KDM5B(75605)

General description

Mouse PLU-1, also known as JARID1B and KDM5B (GenBank Accession No. NM_152895), amino-acids 2-751 with N-terminal FLAG-tag, MW = 86 kDa, expressed in Sf9 cells via a Baculovirus expression system.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kenichi Yamane et al.
Molecular cell, 25(6), 801-812 (2007-03-17)
Posttranslational modification of chromatin by histone methylation has wide-ranging effects on nuclear function, including transcriptional regulation, maintenance of genome integrity, and epigenetic inheritance. The enzymes utilized to place histone methylation marks are well characterized, but the identity of a histone
Angela Barrett et al.
International journal of cancer, 121(2), 265-275 (2007-03-22)
The PLU-1/JARID1B nuclear protein, which is expressed in a high proportion of breast cancers, but shows restricted expression elsewhere, belongs to the ARID family of proteins, known to play important roles in development, differentiation, transcriptional regulation and chromatin remodeling. PLU-1/JARID1B

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service