Skip to Content
Merck
All Photos(2)

Key Documents

SRP0372

Sigma-Aldrich

MKK6 active human

recombinant, expressed in baculovirus infected Sf9 cells, ≥80% (SDS-PAGE)

Synonym(s):

MAP2K6, MEK6, mitogen-activated protein kinase kinase 6

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$726.00

$726.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
20 μG
$726.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

$726.00


Check Cart for Availability

Request a Bulk Order

biological source

human

recombinant

expressed in baculovirus infected Sf9 cells

Assay

≥80% (SDS-PAGE)

form

aqueous solution

mol wt

63.5 kDa

packaging

pkg of 20 μg

storage condition

avoid repeated freeze/thaw cycles

concentration

1.36 mg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... MAP2K6(5608)

General description

Human MKK6 (GenBank Accession No. NM_002758.3), amino acids 2-end with N-terminal GST-tag, MW=63.5 kDa, expressed in a Baculovirus infected Sf9 cell expression system.

Application

Useful for the study of enzyme kinetics, screening inhibitors, and selectivity profiling.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carlo Serra et al.
Molecular cell, 28(2), 200-213 (2007-10-30)
During muscle regeneration, the mechanism integrating environmental cues at the chromatin of muscle progenitors is unknown. We show that inflammation-activated MKK6-p38 and insulin growth factor 1 (IGF1)-induced PI3K/AKT pathways converge on the chromatin of muscle genes to target distinct components
T L Lotan et al.
The Journal of pathology, 212(4), 386-394 (2007-06-20)
Identification of the signalling cascades that are differentially activated during prostatic tumourigenesis is a crucial step in the search for future molecular targets in this disease. The stress-activated protein kinase (SAPK) signalling cascade culminates in the phosphorylation of the JNK

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service