Skip to Content
Merck
All Photos(2)

Key Documents

SRP0362

Sigma-Aldrich

CHK2 active human

recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE)

Synonym(s):

CDS1, LFS2, RAD53, checkpoint kinase 2

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
$760.00

$760.00


Estimated to ship on01 May 2025


Request a Bulk Order

Select a Size

Change View
10 μG
$760.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

$760.00


Estimated to ship on01 May 2025


Request a Bulk Order

biological source

human

recombinant

expressed in baculovirus infected Sf9 cells

Assay

≥70% (SDS-PAGE)

form

aqueous solution

mol wt

91 kDa

packaging

pkg of 10 μg

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... CHEK2(11200)

General description

Human Checkpoint kinase 2 (GenBank Accession No. NM_007194), full length with N-terminal HIS-tag, MW=91kDa, expressed in a Baculovirus infected Sf9 cell expression system.

Application

Useful for the study of enzyme kinetics, screening inhibitors, and selectivity profiling.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Travis H Stracker et al.
Cell cycle (Georgetown, Tex.), 7(23), 3618-3621 (2008-11-26)
Central to the DNA damage response (DDR) is the highly conserved Mre11 complex consisting of Mre11, Rad50 and Nbs1. The Mre11 complex acts as a sensor of DNA double-strand breaks (DSBs) and regulates the signal transduction cascades that are triggered
Reinhardt HC and Yaffe MB.
Current Opinion in Cell Biology (2009)
Qi-Qiang Guo et al.
The EMBO journal, 39(10), e103111-e103111 (2020-03-19)
The homeostatic link between oxidative stress and autophagy plays an important role in cellular responses to a wide variety of physiological and pathological conditions. However, the regulatory pathway and outcomes remain incompletely understood. Here, we show that reactive oxygen species

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service