Skip to Content
Merck
All Photos(1)

Key Documents

SRP0361

Sigma-Aldrich

PLK3 Polo Box Domain human

recombinant, expressed in E. coli, ≥90% (SDS-PAGE)

Synonym(s):

CNK, FNK, PRK, polo-like kinase 3

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$902.00

$902.00


Estimated to ship on22 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$902.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

$902.00


Estimated to ship on22 May 2025


Request a Bulk Order

biological source

human

recombinant

expressed in E. coli

Assay

≥90% (SDS-PAGE)

form

aqueous solution

mol wt

80.5 kDa

packaging

pkg of 100 μg

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... PLK3(1263)

General description

Human N-terminal MBP tagged Plk3 polo box domain (GenBank Accession No.NM_004073/NP_004064), amino acids 335-646 with C-terminal His-tag, MW=80.5 kDa, expressed in an E. coli expression system.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Suqing Xie et al.
Oncogene, 23(21), 3822-3829 (2004-03-17)
MEK1, a gene product that regulates cell growth and differentiation, also plays an important role in Golgi breakdown during the cell cycle. We have recently shown that polo-like kinase (Plk3) is Golgi localized and involved in Golgi dynamics during the
Zhongkui Li et al.
The Journal of biological chemistry, 280(17), 16843-16850 (2005-01-27)
RelA, the p65 subunit of NF-kappaB transcription factors, plays a key role in regulation of antiapoptotic and proapoptotic responses. However, the downstream target genes regulated by RelA-NF-kappaB in the initiation of proapoptotic signaling were not identified. We previously showed that

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service