Skip to Content
Merck
All Photos(2)

Key Documents

SRP0352

Sigma-Aldrich

TRKC active human

recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE)

Synonym(s):

NT-3 Receptor, NTKR3, Neurotrophic Tyrosine Kinase Receptor, type 3

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
$760.00

$760.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 μG
$760.00

About This Item

CAS Number:
UNSPSC Code:
12352200
NACRES:
NA.32

$760.00


Check Cart for Availability

Request a Bulk Order

biological source

human

recombinant

expressed in baculovirus infected Sf9 cells

Assay

≥70% (SDS-PAGE)

form

aqueous solution

mol wt

68 kDa

packaging

pkg of 10 μg

technique(s)

activity assay: suitable
inhibition assay: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... NTRK3(4916)

General description

Human TRKC, also known as NTKR3 (Neurotrophic Tyrosine Kinase Receptor, type 3) or NT-3 Receptor, (GenBank Accession No. NM_002530), amino acids 456-825 (end) with N-terminal GST-tag, MW=68kDa, expressed in Sf9 cells via a Baculovirus expression system.

Application

Useful for the study of enzyme kinetics, screening inhibitors, and selectivity profiling.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L Tessarollo et al.
Development (Cambridge, England), 118(2), 463-475 (1993-06-01)
The Trk family of tyrosine kinases encodes receptors for nerve growth factor-related neurotrophins. Here we present a developmental expression study of trkC, which encodes a receptor for neurotrophin-3 (NT-3). Like the related genes, trk and trkB, trkC is expressed primarily
F Lamballe et al.
Cell, 66(5), 967-979 (1991-09-06)
We report the isolation and molecular characterization of trkC, a new member of the trk family of tyrosine protein kinase genes. trkC is preferentially expressed in the brain. In situ hybridization studies revealed trkC transcripts in the hippocampus, cerebral cortex
L M McGregor et al.
Genomics, 22(2), 267-272 (1994-07-15)
TrkC is a receptor tyrosine kinase that is activated by neurotrophin-3, a factor important in the development of certain areas of the central nervous system. We have cloned and sequenced the human trkC cDNA and found that the predicted amino
Ke Zhang et al.
Stem cell reviews, 10(4), 612-625 (2014-05-03)
In the past decades, mesenchymal stem cells (MSCs) as a promising cell candidate have received the most attention in the treatment of spinal cord injury (SCI). However, due to the low survival rate and low neural differentiation rate, the grafted
Oswald Steward et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 34(42), 14013-14021 (2014-10-17)
We reported previously the formation of ectopic colonies in widespread areas of the nervous system after transplantation of fetal neural stem cells (NSCs) into spinal cord transection sites. Here, we characterize the incidence, distribution, and cellular composition of the colonies.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service