Skip to Content
Merck
All Photos(2)

Key Documents

SRP0341

Sigma-Aldrich

cKit active human

recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE)

Synonym(s):

CD117, KIT, PBT, SCFR, v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene homolog

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
$570.00

$570.00


Estimated to ship on23 May 2025


Request a Bulk Order

Select a Size

Change View
10 μG
$570.00

About This Item

CAS Number:
UNSPSC Code:
12352200
NACRES:
NA.32

$570.00


Estimated to ship on23 May 2025


Request a Bulk Order

biological source

human

recombinant

expressed in baculovirus infected Sf9 cells

Assay

≥70% (SDS-PAGE)

form

aqueous solution

mol wt

53.5 kDa

packaging

pkg of 10 μg

technique(s)

activity assay: suitable
inhibition assay: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... KIT(3815)

General description

Human Kit ( GenBank Accession No. NM_000222), amino acids 544-976 with N-terminal His-tag, MW=50 kDa, expressed in a Baculovirus infected Sf9 cell expression system.

Application

Useful for the study of enzyme kinetics, screening inhibitors, and selectivity profiling.

Unit Definition

One unit is defined as the amount of enzyme that will phosphorylate 1 nmol of tyrosine substrate per minute at pH 7.4 and 30°C.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mauro Picardo et al.
The Journal of investigative dermatology, 131(6), 1182-1185 (2011-05-14)
Cutaneous pigmentation is regulated by a complex melanogenic network in which skin cells synthesize growth factors and cytokines. Mutations in genes encoding these regulators modify their expression and/or functionality, leading to altered signaling pathways and contributing to altered skin phenotypes.
Y Yarden et al.
The EMBO journal, 6(11), 3341-3351 (1987-11-01)
Structural features of v-kit, the oncogene of HZ4 feline sarcoma virus, suggested that this gene arose by transduction and truncation of cellular sequences. Complementary DNA cloning of the human proto-oncogene coding for a receptor tyrosine kinase confirmed this possibility: c-kit

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service