Skip to Content
Merck
All Photos(1)

Key Documents

SRP0254

Sigma-Aldrich

Cofilin 1 Active human

recombinant, expressed in E. coli, ≥90% (SDS-PAGE)

Synonym(s):

CFL1, Cofilin, non-muscle isoform, p18

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

Assay

≥90% (SDS-PAGE)

form

aqueous solution

mol wt

44 kDa

packaging

pkg of 200 μg

concentration

>0.02 mg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... CFL1(1072)

General description

Cofilin 1 is ubiquitously expressed and has a molecular mass of ~19kDa.
Human recombinant Cofilin1 (2-end) with N-terminal GST tag (GenBank Accession No. NM_005507), expressed in a E. coli expression system. Cofilin 1, MW = 44 kDa,
Human recombinant Cofilin1 (2-end) with N-terminal GST tag (GenBank Accession No. NM_005507), expressed in a E. coli expression system. Cofilin 1, MW = 44 kDa,

Application

Kinase substrate for the kinase assay; western blot

Biochem/physiol Actions

Cofilin-l (CFL1) plays a vital role in the regulation of actin cytoskeleton reorganization. It is also implicated in cancer cell migration and invasion in several types of tumor tissues. Elevated expression of the gene is associated with the development of esophageal squamous cell carcinoma (ESCC). In addition, abnormal expression of the gene leads to vulvar cancer and thus, it can be considered as a potential therapeutic target for the same. CFL1 facilitates cytokinesis, cell motility and morphogenesis. Activated CFL1 helps in preventing cytoskeletal-associated defects in axonal development stimulated by Syns.

Physical form

Formulated in 25 mM Tris-HCl, pH 8.0, 100 mM NaCl, 0.05% Tween-20, 30% glycerol and 3 mM DTT.

Preparation Note

Thaw on ice. Upon first thaw, briefly spin tube containing enzyme to recover full content of the tube. Aliquot enzyme into single use aliquots. Store remaining undiluted enzyme in aliquots at -70°C. Note: Enzyme is very sensitive to freeze/thaw cycles.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cofilin 1 activation prevents the defects in axon elongation and guidance induced by extracellular alpha-synuclein.
Tilve S
Scientific Reports (2015)
The role of cofilin-l in vulvar squamous cell carcinoma: A marker of carcinogenesis, progression and targeted therapy.
Wu Q
Oncology Reports, 35, 2743-2754 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service