Skip to Content
Merck
All Photos(1)

Key Documents

SRP0252

Sigma-Aldrich

ULK Active human

recombinant, expressed in FreeStyle 293-F cells, ≥40% (SDS-PAGE)

Synonym(s):

ATG1, ATG1 autophagy related 1 homolog, Serine/threonine-protein kinase ULK1, UNC-51-like kinase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in FreeStyle 293-F cells

Assay

≥40% (SDS-PAGE)

form

aqueous solution

mol wt

113 kDa

packaging

pkg of 10 μg

manufacturer/tradename

Invitrogen

storage condition

avoid repeated freeze/thaw cycles

concentration

>0.02 mg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... ULK1(8408)

General description

ULK1 (unc-51 like autophagy activating kinase 1) is a Ser/Thr protein kinase which forms a component of the ULK1 protein kinase complex. This protein complex also contains the adaptor proteins ATG (autophagy-related) 13, RB1CC1/FIP200 (RB1-inducible coiled-coil 1), and ATG101. ULK1 protein is one of the five orthologs of yeast Atg1 found in mammals.

Biochem/physiol Actions

ULK1 (unc-51 like autophagy activating kinase 1) protein kinase plays a crucial role in the control of autophagy initiation. This protein is responsible for activating a cascade of signaling pathways which ultimately results in the formation of phagosomes. The phosphorylation of ULK1 by MTORC1 (mammalian target of rapamycin complex) leads to the negative regulation of autophagy. Activation of type I interferon (IFN) receptor (IFNR) results in the activation of ULK1 protein, which, in turn, results in the transcription of gene in a IFN-stimulated response elements (ISRE) and IFNγ activation site (GAS) elements-mediated manner. It is also required for the control of the expression of key IFN-stimulated genes (ISGs).

Physical form

Formulated in 2 5mM Tris-HCl, pH 7.5, 100 mM NaCl, 0.05% Tween-20, 100 μg/ml FLAG peptide, 20% glycerol and 3 mM DTT.

Preparation Note

Thaw on ice. Upon first thaw, briefly spin tube containing enzyme to recover full content of the tube. Aliquot enzyme into single use aliquots. Store remaining undiluted enzyme in aliquots at -70°C. Note: Enzyme is very sensitive to freeze/thaw cycles.

Legal Information

FreeStyle is a trademark of Invitrogen Corp.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Deubiquitinase inhibition by WP1130 leads to ULK1 aggregation and blockade of autophagy.
Autophagy, 11(9), 1458-1470 (2015)
Central role of ULK1 in type I interferon signaling.
Saleiro D et al
Cell Reports, 11(4), 605-617 (2015)
Computational analysis of an autophagy/translation switch based on mutual inhibition of MTORC1 and ULK1.
Szymanska P et al
PLoS ONE, 10(3), e0116550-e0116550 (2015)
Pablo Sánchez-Martín et al.
Journal of cell science, 136(3) (2023-01-17)
Autophagy is a catabolic process during which cytosolic material is enwrapped in a newly formed double-membrane structure called the autophagosome, and subsequently targeted for degradation in the lytic compartment of the cell. The fusion of autophagosomes with the lytic compartment

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service