Skip to Content
Merck
All Photos(1)

Documents

SRP0229

Sigma-Aldrich

WWP1 Active human

recombinant, expressed in baculovirus infected insect cells, ≥15% (SDS-PAGE)

Synonym(s):

AIP5, Atrophin-1 interacting protein 5, NEDD4-like E3 ubiquitin-protein ligase, TGIF-interacting ubiquitin ligase 1 (Tiul1), WW domain containing E3 ubiquitin protein ligase 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in baculovirus infected insect cells

Assay

≥15% (SDS-PAGE)

form

aqueous solution

mol wt

106 kDa

packaging

pkg of 20 μg

storage condition

avoid repeated freeze/thaw cycles

concentration

>0.02 mg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... WWP1(11059)

General description

Human WWP1 (GenBank Accession No. NM_007013), amino acids 2-end, with N-terminal DDDDK tag (FLAG), MW = 106 kDa, expressed in a Baculovirus infected Sf9 cell expression system.

Application

Useful in conjunction with E1 and E2 for the study of enzyme kinetics, screening inhibitors, and selectivity profiling.

Unit Definition

One unit is defined as the amount of enzyme required to transfer 1 pmol of ubiquitin to substrate/min at 37°C.

Physical form

Formulated in 25 mM Tris-HCl, pH 8.0, 100 mM NaCl, 0.05% Tween-20, 100 μg/ml FLAG peptide, 20% glycerol and 3 mM DTT.

Preparation Note

Thaw on ice. Upon first thaw, briefly spin tube containing enzyme to recover full content of the tube. Aliquot enzyme into single use aliquots. Store remaining undiluted enzyme in aliquots at -70°C. Note: Enzyme is very sensitive to freeze/thaw cycles.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yu-Ru Lee et al.
Science (New York, N.Y.), 364(6441) (2019-05-18)
Activation of tumor suppressors for the treatment of human cancer has been a long sought, yet elusive, strategy. PTEN is a critical tumor suppressive phosphatase that is active in its dimer configuration at the plasma membrane. Polyubiquitination by the ubiquitin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service