Skip to Content
Merck
All Photos(2)

Key Documents

SRP0189

Sigma-Aldrich

Pyruvate kinase M2 Active from mouse

recombinant, expressed in E. coli, ≥90% (SDS-PAGE)

Synonym(s):

M2-PK tumor, OPA-interacting protein 3 (OIP-3), Pyruvate kinase 3 (PK3), Pyruvate kinase, muscle, Thyroid hormone-binding protein 1 (THBP1)

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$947.00

$947.00


Check Cart for Availability


Select a Size

Change View
20 μG
$947.00

About This Item

UNSPSC Code:
12352204
NACRES:
NA.32

$947.00


Check Cart for Availability

biological source

mouse

recombinant

expressed in E. coli

Assay

≥90% (SDS-PAGE)

form

aqueous solution

mol wt

58 kDa

packaging

pkg of 20 μg

storage condition

avoid repeated freeze/thaw cycles

concentration

>0.02 mg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

mouse ... PKM2(18746)

General description

Pyruvate kinase M2 (PKM2), also known as pyruvate kinase 3 (PK3), is an isoform of pyruvate kinase that is expressed in some differentiated tissues, such as lung, fat, retina, and pancreatic islets, as well as in normal proliferating cells and embryonic cells.[1] PKM2/pk3 is encoded by the gene mapped to mouse chromosome 9.[2]
Murine Pyruvate kinase M2 (PKM2) (GenBank Accession No. NM_011099), amino acids 2-end, with N-terminal His tag, MW = 58kDa, expressed in an Escherichia coli expression system.

Application

Useful for the study of enzyme kinetics, screening inhibitors, and selectivity profiling.

Biochem/physiol Actions

Elevated expression of PKM2 is observed in most of the cancer cells. Hence, this protein can be considered as a potent therapeutic target for cancers.[3][1] PKM2 plays an important role in the discharge of excessive intracellular reactive oxygen species (ROS), which otherwise damage cellular components and decrease cell viability. Protein kinase activity of PKM2 is involved in the regulation of gene expression and tumorigenesis. PKM2 is an important rate-limiting enzyme involved in final step of glycolysis and it has an essential role in metabolic reprogramming throughout cancer progression.[1][3]

Unit Definition

One unit is defined as the amount of enzyme required to transfer a phosphoryl group from phosphoenolpyruvate (PEP) to 1 pmol of ADP/min at 37°C

Physical form

Formulated in 25 mM Tris-HCl, pH 8.0, 100 mM NaCl, 0.05% Tween-20, 30% glycerol and 3 mM DTT.

Preparation Note

Thaw on ice. Upon first thaw, briefly spin tube containing enzyme to recover full content of the tube. Aliquot enzyme into single use aliquots. Store remaining undiluted enzyme in aliquots at -70°C. Note: Enzyme is very sensitive to freeze/thaw cycles.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

PKM2 and cancer: The function of PKM2 beyond glycolysis.
Dong G
Oncology Letters, 11(3), 1980-1986 (2016)
Polymorphism of kidney pyruvate kinase in the mouse is determined by a gene, Pk-3, on chromosome 9.
Peters J
Biochemical Genetics, 19(7-8), 757-769 (1981)
Weiwei Yang et al.
Cancer letters, 339(2), 153-158 (2013-06-25)
Altered metabolism is fundamental to the growth and survival of cancer cells. Pyruvate kinase M2 (PKM2), a key enzyme in cancer metabolism, has been demonstrated to play a central role not only in metabolic reprogramming but also in direct regulation

Articles

Warburg effect enhances glucose to lactate conversion in tumor cells, regardless of oxygen levels; impacting cancer metabolism since 1924.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service