Skip to Content
Merck
All Photos(2)

Key Documents

SRP0169

Sigma-Aldrich

JMJD2B Active human

recombinant, expressed in baculovirus infected insect cells, ≥40% (SDS-PAGE)

Synonym(s):

JHDM3B, JmjC domain-containing histone demethylation protein 3B, Jumonji domain-containing protein 2B, KIAA0876, Lysine-specific demethylase 4B (KDM4B)

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$700.00

$700.00


Estimated to ship on01 June 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$700.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

$700.00


Estimated to ship on01 June 2025


Request a Bulk Order

biological source

human

recombinant

expressed in baculovirus infected insect cells

Assay

≥40% (SDS-PAGE)

form

aqueous solution

mol wt

57 kDa

packaging

pkg of 100 μg

concentration

>0.02 mg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... KDM4B(23030)

General description

Human JMJD2B, also known as JHDM3B and KDM4B (GenBank Accession No.NM_015015) (amino acids 1-500) with C-terminal FLAG tag, MW = 57 kDa, expressed in Sf9 cells using a Baculovirus expression system.

Application

Useful for the study of enzyme kinetics, screening inhibitors, and selectivity profiling.

Unit Definition

One unit is defined as the amount of enzyme required to demethylate 1 pmol of substrate/min at 37°C.

Physical form

Formulated in 25 mM Tris-HCl, pH 8.0, 100 mM NaCl, 0.05% Tween-20, 20% glycerol and 3 mM DTT.

Preparation Note

Thaw on ice. Upon first thaw, briefly spin tube containing enzyme to recover full content of the tube. Aliquot enzyme into single use aliquots. Store remaining undiluted enzyme in aliquots at -70°C. Note: Enzyme is very sensitive to freeze/thaw cycles.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service