Skip to Content
Merck
All Photos(3)

Documents

SRE0115

Sigma-Aldrich

Salt Active Nuclease (SAN)

Heat-labile, recombinant, expressed in Pichia pastoris, Triton-free

Synonym(s):

Heat-labile salt active nuclease, endonuclease, salt active endonuclease, salt tolerant nuclease

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204
NACRES:
NA.54

recombinant

expressed in Pichia pastoris

Quality Level

specific activity

≥175 000 U/mg

feature

Heat-labile

composition

Triton-free

packaging

pkg of 25 kU

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Nucleic acids, and especially genomic DNA, often pose a problem in purification of DNA-binding proteins as they interfere with purification, downstream analysis or applications. As most nucleases are inhibited by high concentrations of salt, removal of DNA is difficult, since NaCl is used to dissociate DNA from proteins.
This Salt Active Nuclease (SAN) is a nonspecific endonuclease recombinantly produced in Pichia pastoris, with optimum activity at high salt concentrations. It is active in a variety of buffers and can be easily inactivated by treatment with a reducing agent. These features make it particularly useful in the purification of proteins and removal of DNA and RNA from molecular biology reagents. It can be used on cell lysates for viscosity removal, and additionally at the elution step of purification, where there are smaller volumes and a purer protein - allowing for effective and efficient DNA removal.
This Salt Active Nuclease (SAN) is a heat-labile nuclease that has optimal activity at high salt concentrations and is useful during protein purification.

Application

The high salt tolerance and easy removal makes this SAN useful in protein purification schemes, especially in combination with IMAC (Immobilized metal affinity chromatography) media used for purification of poly-His-tagged proteins.

Features and Benefits

  • Active at up to 1M NaCl
    Tolerates imidazole and can be separated from most proteins by cation exchange or inactivated by reducing agents.

Unit Definition

One Unit is defined as an increase in absorbance at 260 nm of 1 A in 30 minutes at 37°C, using 50 μg/ml calf thymus DNA (D1501, Sigma) in a buffer consisting of 25 mM Tris-HCl, pH 8.5 (25°C), 5 mM MgCl2, 500 mM NaCl.

Reconstitution

In solution

Other Notes

Temp: 5 – 40°C, optimal: ~ 35°C
Salt concentration (NaCI / KCI): 150 - 900 mM, optimal: 400 - 650 mM
Mg2+: >1 mM is required for activity, optimal: 5 - 50 mM
pH: 7.3 – 10, optimal: 8.2 - 9.2
Not recommended to use in presence of SDS or urea.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service