Skip to Content
Merck
All Photos(7)

Key Documents

SRE0113

Sigma-Aldrich

Bst Max DNA Polymerase

recombinant, expressed in E. coli, Salt and Inhibitor Tolerant

Synonym(s):

Bst DNA polymerase, Isothermal DNA Polymerase

Sign Into View Organizational & Contract Pricing

Select a Size

1600 UNITS
$127.00

$127.00


Check Cart for Availability


Select a Size

Change View
1600 UNITS
$127.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.54

$127.00


Check Cart for Availability

recombinant

expressed in E. coli

Quality Level

specific activity

40,000 U/mg

feature

Salt and Inhibitor Tolerant

optimum pH

8.5

shipped in

dry ice

storage temp.

−20°C

General description

Loop-mediated isothermal amplification (LAMP) has emerged as a key alternative to polymerase chain reaction (PCR)-based techniques in the amplification of nucleic acids. While conventional PCR employs a thermal cycler for repetitive cycling temperatures when amplifying, isothermal amplification techniques such as LAMP occur at a single and fixed temperature allowing the use of simple, portable, and more robust instruments for fast and exponential amplification.
The Bst Max DNA polymerase is an in-silicon designed homolog of Bst DNA Polymerase (large fragment) suitable for DNA amplification at elevated temperatures with an optimum of 65°C. Bst Max DNA polymerase is a salt-tolerant recombinant polymerase (optimal salt concentration 50-350 mM) with more than two-fold enhanced strand-displacement activity and processivity when compared to Bst polymerase. The Bst enzyme is active from 25 to 65°C. It lacks 5’-3’- and 3’-5’-exonuclease activity.

Application

Bst Max DNA Polymerase is
  • Ideal for isothermal applications such as LAMP and RT-LAMP for its superior amplification performance and robustness, especially at point-of-care diagnostics
  • Compatible with a wide range of buffer formulations and isothermal applications such as Ramification amplification (RAM), NEMA (Nicking enyzme-mediated amplification), HAD (Helicase-dependent amplification), MDA/SDA (Multiple/Strand displacement amplification), RCA (Rolling circle amplification), RPA (Recombinase Polymerase amplification)

Features and Benefits

  • Efficient strand displacement activity allows the polymerase to amplify at a faster rate while displacing the second DNA strand during synthesis.
  • High processivity ensures that the polymerase can amplify more nucleotides consecutively without dissociation from the DNA template
  • Higher salt and inhibitor tolerance ensures enhanced amplification performance without the need for sample prep steps
  • Ideal for amplification of most sample types, especially small and impure samples
  • Triton-free formulation complies with the European Chemicals Agency’s REACH regulations to improve the protection of human health and the environment from risks that can be posed by chemicals

Unit Definition

One unit is defined as the amount of enzyme that will incorporate 10 nmol of dNTP into acid insoluble material in 30 minutes at 65°C.

Reconstitution

In solution.

Other Notes

Order primers for LAMP using our online configurator. For assistance with primer design prior to purchase, please use the online design tool from our partner.

related product

Product No.
Description
Pricing

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service