Skip to Content
Merck
All Photos(12)

Key Documents

SRE0093

Sigma-Aldrich

β-Glucuronidase from limpets (Patella vulgata)

aqueous solution

Synonym(s):

β-D-Glucuronide glucuronosohydrolase

Sign Into View Organizational & Contract Pricing

Select a Size

10 ML
$289.00
25 ML
$665.00
50 ML
$1,280.00

$289.00


Check Cart for Availability


Select a Size

Change View
10 ML
$289.00
25 ML
$665.00
50 ML
$1,280.00

About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

$289.00


Check Cart for Availability

recombinant

expressed in proprietary host

Quality Level

form

aqueous solution

concentration

100000-200000 units/mL

shipped in

ambient

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

New Technical Article Comparing Performance of Different Enzymes
Learn more about recent application data generated by Sigma R&D to optimize hydrolysis for different drug classes using enzymes from different sources and the use of a chromatographicaly purified enzyme to reduce the effect of esterase activity resulting in conversion of 6-MAM to Morphine
More on the suitability of this product for hydrolysis of different drug-glucuronide substrates is available at this page.
Poster presentation comparing the hydrolysis efficiency of novel beta-glucuronidase enzymes from recombinant limpet and recombinant E. coli sources, click at the following poster link: Hydrolysis Efficiency Evaluation of Novel Recombinant Limpet and E. coli Beta-Glucuronidase Enzymes
The enzyme from patella vulgata is reported to be more effective in hydrolyzing opioid-glucuronides than the Helix pomatia, bovine liver and E. coli enzymes

Features and Benefits

Recombinantly expressed and purified to eliminate MAM esterase activity (6-Monoacetylmorphine→morphine). Representative analysis at high enzyme titer (50 un/μL β-glucuronidase) was less than 1% conversion of 6-MAM to morphine after 4 hours at 60 °C.

Unit Definition

One Sigma or modified Fishman unit will liberate 1.0 μg of phenolphthalein from phenolphthalein glucuronide per hr at 37 °C at pH 3.8 (30 min assay).

Physical form

Supplied as a solution in 100mM potassium acetate pH 5.2. It is free of detergents, carbohydrates or any other components that may interfere with sample preparation and analysis.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

β-glucuronidase (GUS) enzymes are utilized to hydrolyze glucuronide (gluc) drug metabolites to the parent drug, facilitating analysis by LC-MS/MS.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service