Skip to Content
Merck
All Photos(1)

Key Documents

SRE0065

Sigma-Aldrich

HEPES solution, 1M

Synonym(s):

HEPES Buffer, N-(2-Hydroxyethyl)piperazineN′-(2-ethanesulfonic acid)

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$148.00
500 ML
$458.00
1 L
$788.00

$148.00


In StockDetails


Request a Bulk Order

Select a Size

Change View
100 ML
$148.00
500 ML
$458.00
1 L
$788.00

About This Item

UNSPSC Code:
12352107
NACRES:
NA.25

$148.00


In StockDetails


Request a Bulk Order

form

liquid

Quality Level

concentration

1 M

color

colorless

pH

7.2-7.5 (25 °C)

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

Application

HEPES buffer preserves a normal physiological pH environment and is commonly used as a buffer in cell culture applications. HEPES buffer provides no nutritional benefit to cells, but adds extra buffering capacity to the cell culture particularly during experiments performed outside the carbon dioxide incubator.
This 1 M stock solution contains HEPES adjusted to pH 7.35 with sodium hydroxide. Add the concentrated buffer to the cell culture medium to reach the desired concentration (typically 10–25 mM).

Notes: If precipitation is present in the concentrated buffer, gently warm the bottle to 37 °C and mix until completely dissolved prior to dilution.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service