Skip to Content
Merck
All Photos(1)

Key Documents

SRE0049

Sigma-Aldrich

Lipase from porcine pancreas

Suitable for manufacturing of diagnostic kits and reagents, ≥20,000 units/mg protein, lyophilized powder

Synonym(s):

PPL, Triacylglycerol acylhydrolase, Triacylglycerol lipase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204

Quality Level

form

lyophilized powder

specific activity

≥20,000 units/mg protein

composition

Protein, 40-70% biuret

shipped in

wet ice

storage temp.

−20°C

InChI

1S/C11H9N3O2.Na/c15-8-4-5-9(10(16)7-8)13-14-11-3-1-2-6-12-11;/h1-7,16H,(H,12,14);/q;+1/b13-9-;

InChI key

QWZUIMCIEOCSJF-CHHCPSLASA-N

Looking for similar products? Visit Product Comparison Guide

Application

Lipases are used industrially for the resolution of chiral compounds and the transesterification production of biodiesel.

Biochem/physiol Actions

Lipase is a hydrolytic enzyme produced by various tissues. This enzyme catalyzes the hydrolysis of triacylglycerols to yield glycerol and free fatty acids, which is a rate-limiting for the exclusion of lipoprotein triglycerides from the circulation. It is associated with certain metabolic diseases, including obesity and familial combined hyperlipidemia.[1][2]
Lipases catalyze the hydrolysis of esters in aqueous solutions and the synthesis of esters in non-aqueous solutions. They also produce hydroxyl and carboxylic groups through the hydrolysis of ester linkages in poly(ethylene terephthalate).

Lipases catalyze the hydrolysis of triacylglycerols into glycerol and free fatty acids.

Unit Definition

One unit will hydrolyze 1.0 microequivalent of fatty acid from a triglyceride in 1 hr at pH 7.7 at 37 °C using olive oil.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lipoprotein lipase.
Eckel R H
The New England Journal of Medicine, 320(16), 1060-1068 (1989)
8th International Symposium on Atherosclerosis., 265-267 (1989)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service