SRE0039
Cyanogen Bromide Activated Agarose, High Swell Ratio
lyophilized powder
Synonym(s):
Cyanogen bromide-activated Agarose, CNBr- activated agarose
Select a Size
$107.00
Select a Size
About This Item
$107.00
Recommended Products
biological source
plant
form
lyophilized powder
technique(s)
affinity chromatography: suitable
matrix
cross-linked 4% beaded agarose
matrix spacer
1 atom (when ligands are coupled via isourea derivative or related linkage)
swelling
1 g swells to ≥5 mL
capacity
≥10 mg/mL binding capacity (BSA)(via amino groups)
suitability
suitable for chromatography
application(s)
diagnostic assay manufacturing
shipped in
wet ice
storage temp.
−20°C
Looking for similar products? Visit Product Comparison Guide
General description
Features and Benefits
- Cross-linked 4% beaded agarose matrix for ligand coupling and binding reactions.
- Designed with a 1-atom matrix spacer, facilitating efficient ligand coupling via isourea derivative or related linkage.
- Offers a high binding capacity of at least 10 mg/mL for BSA (via amino groups).
- Ideal for manufacturing diagnostic assays.
Physical form
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
No data available
Flash Point(C)
No data available
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service