Skip to Content
Merck
All Photos(1)

Key Documents

SML3853

Sigma-Aldrich

GSK143 dihydrochloride

≥97% (HPLC)

Synonym(s):

2-(((3R,4R)-3-Aminotetrahydro-2H-pyran-4-yl)amino)-4-(p-tolylamino)pyrimidine-5-carboxamide dihydrochloride, 2-[[(3R,4R)-3-Aminotetrahydro-2H-pyran-4-yl]amino]-4-[(4-methylphenyl)amino]-5-pyrimidinecarboxamide dihydrochloride, GSK 143 dihydrochloride, GSK-143 dihydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$294.00
50 MG
$1,190.00

$294.00


Estimated to ship on14 May 2025


Request a Bulk Order

Select a Size

Change View
10 MG
$294.00
50 MG
$1,190.00

About This Item

Empirical Formula (Hill Notation):
C17H22N6O2·2HCl
CAS Number:
Molecular Weight:
415.32
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.21

$294.00


Estimated to ship on14 May 2025


Request a Bulk Order

Quality Level

Assay

≥97% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: 2 mg/mL, clear (Warmed)

storage temp.

-10 to -25°C

Biochem/physiol Actions

GSK143 is an orally active, potent and selective syk inhibitor (pIC50 = 7.5 or IC50 = 31.6 nM) with good selectivity over 66 protein kinases, including ZAP-70 (pIC50 = 4.7 or IC50 = 20 μM). GSK143 effectively inhibits anti-IgM induced Erk1/2 phosphorylation in Ramos cells (pIC50 = 7.1 or IC50 = 79.4 nM) and demonstrates in vivo efficacy in animal inflammatory disease models, including reverse Arthus passive reaction (10 & 30 mg/kg po. in rats), postoperative ileus (POI, 1, 3 or 10 mg/kg po. in mice), and chronic HDM-induced allergic airways inflammation (30 mg/kg/day po. in mice).
Orally active, potent and highly selective syk inhibitor with in vivo efficacy in animal inflammatory disease models.

Caution

Hygroscopic

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Highly selective SYK inhibitor, GSK143, abrogates survival signals in chronic lymphocytic leukaemia
British Journal of Haematology, 182(6), 927-930 (2018)
Inhibition of spleen tyrosine kinase as treatment of postoperative ileus
Gut, 62(11), 1581-1590 (2013)
Discovery of GSK143, a highly potent, selective and orally efficacious spleen tyrosine kinase inhibitor
Bioorganic & Medicinal Chemistry Letters, 21(20), 6188-6194 (2011)
Syk Regulates Neutrophilic Airway Hyper-Responsiveness in a Chronic Mouse Model of Allergic Airways Inflammation
PLoS ONE, 12(1), e0163614-e0163614 (2017)
Christine Braegelmann et al.
Experimental dermatology, 25(5), 375-379 (2016-02-26)
Spleen tyrosine kinase (SYK) is a protein kinase involved in cell proliferation and the regulation of inflammatory pathways. Due to the increasing evidence that kinase inhibitors have potential as specific anti-inflammatory drugs, we have investigated the potential for SYK inhibition

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service