Skip to Content
Merck
All Photos(1)

Documents

SML3850

Sigma-Aldrich

I-BET432

≥98% (HPLC)

Synonym(s):

(R)-7-((R)-1,2-Dihydroxyethyl)-1,3-dimethyl-5-(1-methyl-1H-pyrazol-4-yl)-1H-benzo[d]azepin-2(3H)-one, GSK 3740432, GSK-3740432, GSK3740432

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H21N3O3
Molecular Weight:
327.38
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

Biochem/physiol Actions

I-BET432 is a bromodomain (BrD) inhibitor with selectivity toward the BET family of bromodomain-containing proteins (BrD1/2 pKd = 8.3/8.1 (BRD2), 8.5/8.1 (BRD3), 8.3/8.2 (BRD4), 8.3/7.8 (BRDT) by DiscoRx BROMOscan) with little or no affinity toward bromodomain-containng proteins (BCPs) and BrDs, including CREBBP, TRIM24, EP300, WDR9 BD2, PBRM1 BD2, TAF1 BD2, BAZ2A (pKd <5.9 from DiscoRx BROMOscan panel). I-BET432 inhibits LPS-stimulated MCP-1 production from human white blood cells (pIC50 = 7.4) with oral availability and good in vivo pharmacokinetic properties.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Identification and Optimization of a Ligand-Efficient Benzoazepinone Bromodomain and Extra Terminal (BET) Family Acetyl-Lysine Mimetic into the Oral Candidate Quality Molecule I-BET432
Journal of Medicinal Chemistry, 65(22), 15174-15207 (2022)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service