Skip to Content
Merck
All Photos(1)

Key Documents

SML3842

Sigma-Aldrich

TAT-SAMβA trifluoroacetate

≥95% (HPLC)

Synonym(s):

SAMβA trifluoroacetate, TAT(47-57)-GG-βIIPKC(624-632) trifluoroacetate, TAT(47-57)-GG-SAMβA trifluoroacetate, YGRKKRRQRRR-GG-RNAENFDRF trifluoroacetate

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$360.00

$360.00


Estimated to ship on31 May 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$360.00

About This Item

Empirical Formula (Hill Notation):
C118H195N51O31 · xC2HF3O2
Molecular Weight:
2824.13 (free base basis)
UNSPSC Code:
12352200
NACRES:
NA.77

$360.00


Estimated to ship on31 May 2025


Request a Bulk Order

Quality Level

Assay

≥95% (HPLC)

form

(Powder or Lyophilized powder or film)

storage condition

desiccated

color

white to off-white

storage temp.

-10 to -25°C

Biochem/physiol Actions

Mitofusin 1 (MFN1)-βIIPKC association blocking peptide that prevents βIIPKC-mediated negative regulation of MFN1.
TAT-SAMβA is a mitofusin 1 (MFN1)-βIIPKC association blocking peptide that prevents βIIPKC-mediated negative regulation of MFN1. TAT-SAMβA protects neonatal and adult rat cardiac myocytes, but not Mfn1-knockout cells, from stress-induced death in cultures and improves cardiac contractility in rats with heart failure in vivo (3 mg/kg/day for 6 wks via Alzet pump delivery) by re-establishing mitochondrial morphology and function. TAT-SAMβA is composed of a 9-aa βIIPKC-derived MFN1-interacting sequence linked at the N-terminal to the cell permeable TAT(47-57) via a -Gly-Gly- spacer

Caution

Hygroscopic

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Mfn1-?IIPKC Interaction Regulates Mitochondrial Dysfunction via Sirt3 Following Experimental Subarachnoid Hemorrhage
Translational Stroke Research, 13(5), 845-857 (2022)
A selective inhibitor of mitofusin 1-?IIPKC association improves heart failure outcome in rats
Nature Communications, 10(1), 329-329 (2019)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service