Skip to Content
Merck
All Photos(2)

Documents

SML3840

Sigma-Aldrich

Phenformin hydrochloride

≥98% (HPLC)

Synonym(s):

1-(Diaminomethylidene)-2-(2-phenylethyl)guanidine hydrochloride, 1-Carbamimidamido-N-(2-phenylethyl)methanimidamide hydrochloride, 1-Phenethyl-biguanide hydrochloride, N-(2-Phenylethyl)imidodicarbonimidic diamide hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H15N5·HCl
CAS Number:
Molecular Weight:
241.72
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.21

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: 2 mg/mL, clear

storage temp.

-10 to -25°C

Biochem/physiol Actions

Phenformin is an orally available biguanide hypoglycemic drug. Phenformin can cause lactic acidosis thus it was withdrawn from most markets. Phenformin exhibits potent anticancer activity associated with AMPK activation and suppression of mTOR phosphorylation.

Caution

Hygroscopic

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rattanaporn Jaidee et al.
Naunyn-Schmiedeberg's archives of pharmacology, 393(9), 1681-1690 (2020-05-10)
Phenformin (Phen), a potent activator of AMPK, is effective against some resistant cancers. This study evaluated the inhibition of proliferation, migration, invasion, and angiogenesis by Phen in aggressive cancer cells and investigated the underlying mechanism of the inhibition. Cholangiocarcinoma (CCA)
A review of phenformin, metformin, and imeglimin
Drug Development Research, 81(4), 390-401 (2020)
Guanyi Liu et al.
Cells, 11(15) (2022-08-13)
The treatment of many skin inflammation diseases, such as psoriasis and atopic dermatitis, is still a challenge and inflammation plays important roles in multiple stages of skin tumor development, including initiation, promotion and metastasis. Phenformin, a biguanide drug, has been

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service