SML3831
22-NBD Cholesterol

≥98% (HPLC)
Synonym(s):
(3β,20S)-20-Methyl-21-[(7-nitro-2,1,3-benzoxadiazol-4-yl)amino]pregn-5-en-3-ol, 22-(N-(7-Nitrobenz-2-oxa-1,3-diazol-4-yl)amino)-23,24-bisnor-5-cholen-3β-ol, 22-(N-(NBD)amino)-23,24-bisnorcholesterol, 22-NBD-Chol, Fluoresterol
About This Item
Quality Level
Assay
≥98% (HPLC)
form
powder
color
, Yellow to dark orange
solubility
DMSO: 2 mg/mL, clear
storage temp.
-10 to -25°C
SMILES string
C[C@]12[C@]([H])([C@]3([C@@]([H])([C@]4(C)C(C[C@@H](O)CC4)=CC3)CC2)[H])CC[C@@]1([C@@H](CNC(C5=NON=C56)=CC=C6[N+]([O-])=O)C)[H]
InChI
1S/C28H38N4O4/c1-16(15-29-23-8-9-24(32(34)35)26-25(23)30-36-31-26)20-6-7-21-19-5-4-17-14-18(33)10-12-27(17,2)22(19)11-13-28(20,21)3/h4,8-9,16,18-22,29,33H,5-7,10-15H2,1-3H3/t16-,18+,19+,20-,21+,22+,27+,28-/m1/s1
InChI key
DLWLXTLRGQWGPC-XIWKFBMMSA-N
Biochem/physiol Actions
22-NBD Cholesterol (22-NBD-Cho), a fluorescence-labeled cholesterol, is a lipid microenvironment-sensitive dye and localizes in the membrane’s interior. 22-NBD Cholesterol has proven useful for real-time imaging of HDL-mediated cholesterol uptake, efflux, and intracellular trafficking in living cells. 22-NBD-cholesterol′s fluorescence intensity is reported to enhance in response to phase changes of membrane lipids from rafts to non-raft fractions, and orient opposite to cholesterol in lipid bilayers. 22-NBD-Chol serves as a fluorescent substrate for cholesterol-converting oxidoreductases. λ ex/em ~ 470-490/500-600 nm (the fluorescence intensity and emission maximum depend on the medium’s polarity).
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service