Skip to Content
Merck
All Photos(2)

Key Documents

SML3830

Sigma-Aldrich

BAF312

≥98% (HPLC)

Synonym(s):

(E)-1-(4-(1-(((4-cyclohexyl-3-(trifluoromethyl)benzyl)oxy)imino)ethyl)-2-ethylbenzyl)azetidine-3-carboxylic acid, 1-[[4-[(1E)-1-[[[4-Cyclohexyl-3-(trifluoromethyl)phenyl]methoxy]imino]ethyl]-2-ethylphenyl]methyl]-3-azetidinecarboxylic acid, 1-{4-[(1E)-(4-Cyclohexyl-3-trifluoromethyl-benzyloxyimino)-ethyl]-2-ethyl-benzyl}-azetidine-3-carboxylic acid, BAF 312, BAF-312, Siponimod

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$163.00
50 MG
$526.00

$163.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
$163.00
50 MG
$526.00

About This Item

Empirical Formula (Hill Notation):
C29H35F3N2O3
CAS Number:
Molecular Weight:
516.60
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$163.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

FC(F)(F)c1c(ccc(c1)CONC(=C)c3cc(c(cc3)CN4CC(C4)C(=O)O)CC)C2CCCCC2

InChI

1S/C29H35F3N2O3/c1-3-21-14-23(10-11-24(21)15-34-16-25(17-34)28(35)36)19(2)33-37-18-20-9-12-26(22-7-5-4-6-8-22)27(13-20)29(30,31)32/h9-14,22,25,33H,2-8,15-18H2,1H3,(H,35,36)

InChI key

VVEHSNDBXFCTMJ-UHFFFAOYSA-N

Biochem/physiol Actions

BAF312 (Siponimod) is an orally active, potent and subtype-selective sphingosine-1-phosphate (S1P) receptor agonist (GTPγ binding EC50 = 0.39 nM/S1P1, 0.98 nM/S1P5, 750 nM/S1P4, >1 µM/S1P3, >10 µM/S1P2). BAF312 oral administration causes a dose-dependent decrease in peripheral lymphocyte counts (by 88% 8h post 1 mg/kg, by 50% 6h post 0.14 mg/kg).
Orally active, potent and subtype-selective sphingosine-1-phosphate (S1P) receptor S1P1 & S1P5 agonist.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Gergely et al.
British journal of pharmacology, 167(5), 1035-1047 (2012-06-01)
BAF312 is a next-generation sphingosine 1-phosphate (S1P) receptor modulator, selective for S1P(1) and S1P(5 ) receptors. S1P(1) receptors are essential for lymphocyte egress from lymph nodes and a drug target in immune-mediated diseases. Here, we have characterized the immunomodulatory potential
Samuel J Jackson et al.
Journal of neuroinflammation, 8, 76-76 (2011-07-07)
Microglial activation in multiple sclerosis has been postulated to contribute to long-term neurodegeneration during disease. Fingolimod has been shown to impact on the relapsing remitting phase of disease by modulating autoreactive T-cell egress from lymph organs. In addition, it is
Discovery of BAF312 (Siponimod), a Potent and Selective S1P Receptor Modulator
ACS Medicinal Chemistry Letters, 4(3), 333-337 (2013)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service