Skip to Content
Merck
All Photos(1)

Documents

SML3810

Sigma-Aldrich

p-Nitrobenzyl mesylate

≥98% (HPLC)

Synonym(s):

4-Nitro-benzenemethanol Methanesulfonate, 4-Nitrobenzyl Mesylate, PNBM

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H9NO5S
CAS Number:
Molecular Weight:
231.23
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.21

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

Biochem/physiol Actions

p-Nitrobenzyl mesylate (PNBM) is a thiol-specific alkylating reagent for direct kinase substrate identification. p-Nitrobenzyl mesylate (PNBM) alkylates thiophosphorylated proteins to form thiophosphate ester epitopes that can be detected by thiophosphate-ester-specific antibody by immunoblotting, immunoprecipitation or immunoaffinity purification.

Caution

Hygroscopic

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

PKC?/? inhibition activates an ULK2-mediated interferon response to repress tumorigenesis
Molecular Cell, 81(21), 4509-4526 (2021)
A semisynthetic epitope for kinase substrates
Nature Methods, 4(6), 511-516 (2007)
Identification of AMPK Phosphorylation Sites Reveals a Network of Proteins Involved in Cell Invasion and Facilitates Large-Scale Substrate Prediction
Cell Metabolism, 22(5), 907-921 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service