Skip to Content
Merck
All Photos(2)

Key Documents

SML3792

Sigma-Aldrich

MRIA9 Trifluoroacetate

≥96% (HPLC)

Synonym(s):

8-(((2r,5r)-5-Amino-1,3-dioxan-2-yl)methyl)-6-(2-chloro-4-(3-fluoropyridin-2-yl)phenyl)-2-(methylamino)pyrido[2,3-d]pyrimidin-7(8H)-one trifluoroacetate, 8-[(trans -5-Amino-1,3-dioxan-2-yl)methyl]-6-[2-chloro-4-(3-fluoro-2-pyridinyl)phenyl]-2-(methylamino)pyrido[2,3-d]pyrimidin-7(8H)-one trifluoroacetate

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$311.00
25 MG
$1,250.00

$311.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$311.00
25 MG
$1,250.00

About This Item

Empirical Formula (Hill Notation):
C24H22ClFN6O3 · xC2HF3O2
Molecular Weight:
496.92 (free base basis)
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.21

$311.00


Estimated to ship on28 May 2025


Request a Bulk Order

Quality Level

Assay

≥96% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

Fc1c(nccc1)c2cc(c(cc2)C3=Cc4c(nc(nc4)NC)N(C3=O)C[C@@H]5OC[C@H](CO5)N)Cl

InChI key

QKNBRNSGPNCARD-SGNKCFNYSA-N

Biochem/physiol Actions

ATP site-targeting, dual salt-inducible kinase (SIK1/2/3) & p21-activated kinase (PAK2/3) inhibitor with excellent selectivity over other kinases.
MRIA9 is an ATP site-targeting, dual salt-inducible kinase (SIK1/2/3 IC50 = 55/48/22 nM; SIK2/3 IC50 = 180/127 nM by cell-based NanoBRET) & p21-activated kinase (PAK2/3 IC50 = 41/140 nM; PAK1 IC50 = 580 nM) inhibitor with excellent selectivity over other kinases. MRIA9 sensitizes ovarian cancer cells to mitotic agent paclitaxel (Pac) treatment in SKOV3 cultures (1 nM Pac plus 0.5-5 µM MRIA9) and HeLa spheroids (2 nM Pac plus 5 µM MRIA9).

Caution

Hygroscopic

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Roberta Tesch et al.
Journal of medicinal chemistry, 64(12), 8142-8160 (2021-06-05)
Salt-inducible kinases (SIKs) are key metabolic regulators. The imbalance in SIK function is associated with the development of diverse cancers, including breast, gastric, and ovarian cancers. Chemical tools to clarify the roles of SIK in different diseases are, however, sparse

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service