Skip to Content
Merck
All Photos(1)

Key Documents

SML3786

Sigma-Aldrich

Polaprezinc

≥97% (Zinc Elemental Analysis)

Synonym(s):

(N-(3-aminopropionyl)-L-histidinato zinc, N-β-Alanyl- L-histidine zinc complex, Zinc L-carnosine, beta-alanyl-L-histidinato zinc

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$97.20
50 MG
$394.00

$97.20


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
$97.20
50 MG
$394.00

About This Item

Empirical Formula (Hill Notation):
C9H12N4O3Zn
CAS Number:
Molecular Weight:
289.61
UNSPSC Code:
12352200
NACRES:
NA.21

$97.20


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥97% (Zinc Elemental Analysis)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

[Zn]1N([C@H](C(=O)O1)Cc2[nH]cnc2)C(=O)CCN

InChI key

IGXZLYMCFZHNKW-FJXQXJEOSA-L

Biochem/physiol Actions

Mucosal protective agent that exhibits ROS-quenching activities; stimulates Hsp70 expression
Polaprezinc is an orally available Zinc L-carnosine complex. It is a mucosal protective agent that exhibits ROS-quenching activities, which promotes the repair of damaged tissues. Polaprezinc exerts cytoprotection through stimulation of Hsp70 expression that prevents the development of inflammatory process in the large intestinal mucosa. It is approved in Japan and Korea for treatment of gastric ulcers.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Role of nutritional zinc in the prevention of osteoporosis
Molecular and Cellular Biochemistry, 338(1-2), 241-254 (2010)
Antioxidant, Anti-inflammatory, and Genomic Stability Enhancement Effects of Zinc l-carnosine: A Potential Cancer Chemopreventive Agent
Nutrition and Cancer, 69(2), 201-210 (2017)
Drug repositioning of polaprezinc for bone fracture healing
Communications biology, 5(1), 462-462 (2022)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service