Skip to Content
Merck
All Photos(2)

Key Documents

SML3785

Sigma-Aldrich

Levonadifloxacin

≥98% (HPLC)

Synonym(s):

(5S)-9-Fluoro-6,7-dihydro-8-(4-hydroxy-1-piperidinyl)-5-methyl-1-oxo-1H,5H-benzo[ij]quinolizine-2-carboxylic acid, (S)-(-)-Nadifloxacin, (S)-9-Fluoro-8-(4-hydroxypiperidin-1-yl)-5-methyl-1-oxo-6,7-dihydro-1H,5H-pyrido[3,2,1-ij]quinoline-2-carboxylic acid, WCK 771

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$163.00
50 MG
$657.00

$163.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
$163.00
50 MG
$657.00

About This Item

Empirical Formula (Hill Notation):
C19H21FN2O4
CAS Number:
Molecular Weight:
360.38
MDL number:
UNSPSC Code:
51111800
UNSPSC Code:
12352200
NACRES:
NA.21

$163.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

Biochem/physiol Actions

Levonadifloxacin is a potent benzoquinolizine fluoroquinolone antibiotic that is affective against Gram-positive and Gram-negative bacteria, including methicillin- and quinolone-resistant Staphylococcus aureus, Streptococcus pneumoniae, Streptococcus pyogenes, Haemophilus influenzae, and Moraxella catarrhalis and atypical pathogens. It appears that Levonadifloxacin targets staphylococcal DNA gyrase, unlike other quinolones, which primarily inhibit DNA topoisomerase IV.
Potent benzoquinolizine fluoroquinolone antibiotic that is affective against Gram-positive and Gram-negative bacteria

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Levonadifloxacin, a Novel Broad-Spectrum Anti-MRSA Benzoquinolizine Quinolone Agent: Review of Current Evidence
Drug design, development and therapy, 13, 4351-4365 (2019)
In vitro bactericidal activity of levonadifloxacin (WCK 771) against methicillin- and quinolone-resistant Staphylococcus aureus biofilms
Journal of Medical Microbiology, 68(8), 1129-1136 (2019)
Structural Characterization of the Millennial Antibacterial (Fluoro)Quinolones-Shaping the Fifth Generation
Pharmaceutics, 13(8), 1289-1289 (2021)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service