Skip to Content
Merck
All Photos(1)

Key Documents

SML3775

Sigma-Aldrich

Desthiobiotin-Phenol

≥98% (HPLC)

Synonym(s):

(4R,5S)-N-[2-(4-Hydroxyphenyl)ethyl]-5-methyl-2-oxo-4-imidazolidinehexanamide, DBP, Desthiobiotin-Tyramide, N-(4-Hydroxyphenethyl)-6-((4R,5S)-5-methyl-2-oxoimidazolidin-4-yl)hexanamide, d-Desthiobiotin-Phenol

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$196.00
25 MG
$822.00

$196.00


Estimated to ship on01 June 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$196.00
25 MG
$822.00

About This Item

Empirical Formula (Hill Notation):
C18H27N3O3
CAS Number:
Molecular Weight:
333.43
UNSPSC Code:
12352200
NACRES:
NA.21

$196.00


Estimated to ship on01 June 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

Biochem/physiol Actions

A peroxidase-sensitive desthiobiotin-phenol (DBP) probe that offers more efficient recovery of DBP-labeled peptides from streptavidin beads and stronger mass spectrometry intensity of modified membrane proteins and achieves greater sensitivity for tyrosine residues detection. DBP shows a similar reactivity to the protein when it is converted to the phenoxyl radical state as biotin-phenol/biotin-tyramide (BP) in ascorbate peroxidase (APEX) proximity labeling. DBP exhibits reduced affinity to streptavidin and does not suffer from sulfur oxidation issue during the radical generation.
APEX (ascorbate peroxidase) proximity tyrosine site label, a sulfur-free Biotin-Tyramide analog that can be displaced competitively by biotin

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N'-acyl-N-methylphenylenediamine as a novel proximity labeling agent for signal amplification in immunohistochemistry
Bioorganic & Medicinal Chemistry, 27(6), 1110-1118 (2019)
Enzymatic Recording of Local Hydrogen Peroxide Generation Using Genetically Encodable Enzyme
Analytical Chemistry, 14869-14877, 94(43)-94(43) (2022)
Jiyoon Beon et al.
eLife, 11 (2022-05-14)
Inositol polyphosphate multikinase (IPMK), a key enzyme in inositol polyphosphate (IP) metabolism, is a pleiotropic signaling factor involved in major biological events, including transcriptional control. In the yeast, IPMK and its IP products promote the activity of the chromatin remodeling

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service