Skip to Content
Merck
All Photos(1)

Documents

SML3758

Sigma-Aldrich

Trpvicin

≥98% (HPLC)

Synonym(s):

N-(5-(2-(2-Cyanopropan-2-yl)pyridin-4-yl)-4-(trifluoromethyl)thiazol-2-yl)-4,6-dimethoxypyrimidine-5-carboxamide, N-[5-[2-(1-Cyano-1-methylethyl)-4-pyridinyl]-4-(trifluoromethyl)-2-thiazolyl]-4,6-dimethoxy-5-pyrimidinecarboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H17F3N6O3S
CAS Number:
Molecular Weight:
478.45
UNSPSC Code:
12352200
NACRES:
NA.21

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

Biochem/physiol Actions

Trpvicin is a sub-type selective and potent inhibitor of TRPV3. Trpvicin shows efficacy in the inhibition of itch and hair loss in mouse models. It appears that Trpvicin stabilizes TRPV3 channel in its a closed state.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Structural basis of TRPV3 inhibition by an antagonist
Nature Chemical Biology, 19(1), 81-90 (2023)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service