Skip to Content
Merck
All Photos(2)

Key Documents

SML3718

Sigma-Aldrich

SIAIS630121

≥98% (HPLC)

Synonym(s):

(E)-N-(4-(1-(4-(4-(8-(2-(2,6-dioxopiperidin-3-yl)-1-oxoisoindolin-4-yl)oct-7-yn-1-yl)piperazin-1-yl)benzoyl)piperidin-4-yl)butyl)-3-(pyridin-3-yl)acrylamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$213.00
25 MG
$854.00

$213.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$213.00
25 MG
$854.00

About This Item

Empirical Formula (Hill Notation):
C49H59N7O5
Molecular Weight:
826.04
UNSPSC Code:
51111800
UNSPSC Code:
12352200
NACRES:
NA.21

$213.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (Warmed)

storage temp.

-10 to -25°C

SMILES string

O=C(/C=C/C1=CN=CC=C1)NCCCCC2CCN(CC2)C(C3=CC=C(C=C3)N4CCN(CC4)CCCCCCC#CC5=CC=CC6=C5CN(C6=O)C7C(NC(CC7)=O)=O)=O

InChI

1S/C49H59N7O5/c57-45(22-16-38-12-10-26-50-35-38)51-27-7-6-11-37-24-29-55(30-25-37)48(60)40-17-19-41(20-18-40)54-33-31-53(32-34-54)28-8-4-2-1-3-5-13-39-14-9-15-42-43(39)36-56(49(42)61)44-21-23-46(58)52-47(44)59/h9-10,12,14-20,22,26,35,37,44H,1-4,6-8,11,21,

InChI key

JTTGVFCANBFLCE-CJLVFECKSA-N

Biochem/physiol Actions

Orally available, cell penetrant, potent and highly selective PROTAC that induces NAMPT degradation in several cancer cells lines
SIAIS630121 is an orally available, cell penetrant, potent and highly selective proteolysis-targeting chimera (PROTAC) that induces NAMPT (nicotinamide phosphoribosyltransferase) degradation in several cancer cells lines (SW620, HT29, and HCT116). It contains NAMPT inhibitor FK866 linked to cereblon (CRBN) ligand lenalidomide. SIAIS630121 exhibits rapid and potent degradation of iNAMPT and eNAMPT.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Addressing the Enzyme-independent tumor-promoting function of NAMPT via PROTAC-mediated degradation
Cell Chemical Biology, 29(11), 1616-1629 (2022)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service