Skip to Content
Merck
All Photos(2)

Key Documents

SML3716

Sigma-Aldrich

MS6105

≥98% (HPLC)

Synonym(s):

2-(5-(Cyclopropylmethyl)-3-(3-(6-(13-(((S)-1-((2S,4R)-4-hydroxy-2-((4-(4-methylthiazol-5-yl)benzyl)carbamoyl)pyrrolidin-1-yl)-3,3-dimethyl-1-oxobutan-2-yl)amino)-13-oxotridecanamido)hex-1-yn-1-yl)phenyl)-4-(4-sulfamoylbenzyl)-1H-pyrazol-1-yl)thiazole-4-carboxylic acid

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$294.00
25 MG
$1,190.00

$294.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$294.00
25 MG
$1,190.00

About This Item

Empirical Formula (Hill Notation):
C65H81N9O9S3
CAS Number:
Molecular Weight:
1228.59
UNSPSC Code:
12352200
NACRES:
NA.77

$294.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (Warmed)

storage temp.

-10 to -25°C

SMILES string

O=C(NCCCCC#CC1=CC(C2=NN(C(CC3CC3)=C2CC4=CC=C(C=C4)S(N)(=O)=O)C5=NC(C(O)=O)=CS5)=CC=C1)CCCCCCCCCCCC(N[C@H](C(N6C[C@@H](C[C@H]6C(NCC7=CC=C(C=C7)C8=C(N=CS8)C)=O)O)=O)C(C)(C)C)=O

InChI

1S/C65H81N9O9S3/c1-43-59(85-42-69-43)48-30-26-47(27-31-48)39-68-61(78)55-38-50(75)40-73(55)62(79)60(65(2,3)4)71-57(77)23-16-11-9-7-5-6-8-10-15-22-56(76)67-34-17-13-12-14-19-44-20-18-21-49(35-44)58-52(36-45-28-32-51(33-29-45)86(66,82)83)54(37-46-24-25-46)7

InChI key

APEYSKGQWJKJAZ-LCLGPXMOSA-N

Biochem/physiol Actions

Cell penetrant, potent and selective lactate dehydrogenase (LDH) degrader (PROTAC) that effectively degrades LDHA and LDHB in PANC1 cells
MS6105 is a cell penetrant, potent and selective lactate dehydrogenase (LDH) degrader (PROTAC) that effectively degrades LDHA and LDHB in multiple pancreatic cancer cell lines. MS6105 potently inhibits the proliferation of PANC1 cells.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ning Sun et al.
Journal of medicinal chemistry, 66(1), 596-610 (2022-12-21)
Lactate dehydrogenase (LDH) is a key glycolytic enzyme and biomarker of aggressive cancers. LDHA and LDHB are two main LDH subunits, and both are frequently overexpressed in tumors and essential for tumor growth. A number of LDHA/B small-molecule inhibitors have

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service