Skip to Content
Merck
All Photos(1)

Key Documents

SML3689

Sigma-Aldrich

S-Geranylgeranyl-L-glutathione

≥98% (HPLC)

Synonym(s):

(S)-2-Amino-5-((R)-1-(carboxymethylamino)-1-oxo-3-((2E,6E,10E)-3,7,11,15-tetramethylhexadeca-2,6,10,14-tetraenylthio)propan-2-ylamino)-5-oxopentanoic acid, GGG

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$163.00
25 MG
$657.00

$163.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$163.00
25 MG
$657.00

About This Item

Empirical Formula (Hill Notation):
C30H49N3O6S
CAS Number:
Molecular Weight:
579.79
UNSPSC Code:
12352200
NACRES:
NA.21

$163.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (Warmed)

storage temp.

2-8°C

SMILES string

OC([C@@H](N)CCC(N[C@H](C(NCC(O)=O)=O)CSC/C=C(C)/CC/C=C(C)/CC/C=C(C)/CCC=C(C)C)=O)=O

InChI

1S/C30H49N3O6S/c1-21(2)9-6-10-22(3)11-7-12-23(4)13-8-14-24(5)17-18-40-20-26(29(37)32-19-28(35)36)33-27(34)16-15-25(31)30(38)39/h9,11,13,17,25-26H,6-8,10,12,14-16,18-20,31H2,1-5H3,(H,32,37)(H,33,34)(H,35,36)(H,38,39)/b22-11+,23-13+,24-17+/t25-,26-/m0/s1

InChI key

BAJIBEVLRWVBAZ-DPRMOSMCSA-N

Biochem/physiol Actions

Potent and selective ligand for P2RY8
The metabolite S-Geranylgeranyl-L-glutathione (GGG) is a potent and selective ligand for P2RY8, a Gα13-coupled receptor. GGG promotes internalization of P2RY8 and inhibits the migration of P2RY8-expressing cells (10 – 100 nM range in WEHI-231 cells). Extracellular GGG is shown to be actively transported by ABCC1 (ATP-binding cassette subfamily C member 1) and catabolized by GGT5 (gamma-glutamyltransferase-5) to S-geranylgeranyl-L-Cys-Gly plus glutathione.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service