Skip to Content
Merck
All Photos(1)

Key Documents

SML3655

Sigma-Aldrich

LockdownPro

≥97% (HPLC)

Synonym(s):

Tetraethyl 4,4′-((propane-2,2-diylbis(4,1-phenylene))bis(oxy))diphthalate

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$159.00
25 MG
$640.00

$159.00


Estimated to ship on25 May 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$159.00
25 MG
$640.00

About This Item

Empirical Formula (Hill Notation):
C39H40O10
Molecular Weight:
668.73
UNSPSC Code:
12352200
NACRES:
NA.25

$159.00


Estimated to ship on25 May 2025


Request a Bulk Order

Quality Level

Assay

≥97% (HPLC)

form

(oil or liquid)

color

colorless to yellow

storage temp.

2-8°C

SMILES string

CC(C1=CC=C(C=C1)OC2=CC(C(OCC)=O)=C(C(OCC)=O)C=C2)(C3=CC=C(OC4=CC(C(OCC)=O)=C(C(OCC)=O)C=C4)C=C3)C

InChI

1S/C39H40O10/c1-7-44-35(40)31-21-19-29(23-33(31)37(42)46-9-3)48-27-15-11-25(12-16-27)39(5,6)26-13-17-28(18-14-26)49-30-20-22-32(36(41)45-8-2)34(24-30)38(43)47-10-4/h11-24H,7-10H2,1-6H3

InChI key

ROEOSYBNNXMANC-UHFFFAOYSA-N

Biochem/physiol Actions

Cell permeable, potent, and selective allosteric inhibitor of phosphatase PPM1F that inhibits tissue invasion by PPM1F-overexpressing human cancer cells
LockdownPro, an esterified lockdown analog with better cell permeability, is a cell permeable, potent, and selective allosteric inhibitor of phosphatase PPM1F. LockdownPro potently inhibits tissue invasion by PPM1F-overexpressing human cancer cells. It does not inhibit PPM1F in vitro.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lockdown, a selective small-molecule inhibitor of the integrin phosphatase PPM1F, blocks cancer cell invasion
Cell Chemical Biology, 29(6), 930-946 (2022)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service