Skip to Content
Merck
All Photos(1)

Key Documents

SML3630

Sigma-Aldrich

MK-0159

≥98% (HPLC)

Synonym(s):

MK 0159, MK0159, N-((1r,4r)-4-(2-Methoxyethoxy)cyclohexyl)-6-(thiazol-5-yl)-1H-pyrrolo[2,3-b]pyridine-4-carboxamide, N-[trans-4-(2-Methoxyethoxy)cyclohexyl]-6-(5-thiazolyl)-1H-pyrrolo[2,3-b]pyridine-4-carboxamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$177.00
25 MG
$712.00

$177.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$177.00
25 MG
$712.00

About This Item

Empirical Formula (Hill Notation):
C20H24N4O3S
CAS Number:
Molecular Weight:
400.49
UNSPSC Code:
12352200
NACRES:
NA.77

$177.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(C1=C2C(NC=C2)=NC(C3=CN=CS3)=C1)N[C@H]4CC[C@@H](CC4)OCCOC

InChI key

JUVJFMVGYBBDKN-HDJSIYSDSA-N

Biochem/physiol Actions

MK-0159 is an orally active, potent and selective CD38 inhibitor (h/m/r CD38 IC50 = 22/3/70 nM) that increases extra- and intracellular NAD+ levels in high CD38-expressing human epithelial A549 cultures, as well as intracellular NAD+/NADP+ in HMVECs (0.3-50 µM). MK-0159 (3-30 mg/kg p.o.) increases systemic NAD+ and decreases ADPR levels in mice, providing protection from myocardial damage upon cardiac I/R injury in vivo (30 mg/kg p.o.).
Potent and selective CD38 inhibitor that protects mice from myocardial damage upon cardiac I/R injury in vivo.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ping-Min Chen et al.
Science advances, 8(24), eabo4271-eabo4271 (2022-06-16)
Infection is one of the major causes of mortality in patients with systemic lupus erythematosus (SLE). We previously found that CD38, an ectoenzyme that regulates the production of NAD+, is up-regulated in CD8+ T cells of SLE patients and correlates
Bharat Lagu et al.
Journal of medicinal chemistry, 65(13), 9418-9446 (2022-06-29)
CD38 is one of the major nicotinamide adenine dinucleotide (NAD+)- and nicotinamide adenine dinucleotide phosphate (NADP+)-consuming enzymes in mammals. NAD+, NADP+, and their reduced counterparts are essential coenzymes for numerous enzymatic reactions, including the maintenance of cellular and mitochondrial redox

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service