Skip to Content
Merck
All Photos(2)

Key Documents

SML3608

Sigma-Aldrich

Massonianoside B

≥95% (HPLC)

Synonym(s):

4-[(2S,3R)-2,3-Dihydro-7-hydroxy-3-(hydroxymethyl)-5-(3-hydroxypropyl)-2-benzofuranyl]-2-methoxyphenyl 6-deoxy-α-L-mannopyranoside

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$327.00
25 MG
$1,310.00

$327.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$327.00
25 MG
$1,310.00

About This Item

Empirical Formula (Hill Notation):
C25H32O10
CAS Number:
Molecular Weight:
492.52
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.21

$327.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥95% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

Biochem/physiol Actions

Cell-permeable, potent, highly selective, reversible and SAM-competitive DOT1L inhibitor
Massonianoside B is a cell-permeable, potent, highly selective, reversible and SAM-competitive DOT1L (disruptor of 4 telomeric silencing 1-like) inhibitor that decreases cellular levels of H3K79 methylation and inhibits the MLL fusion target genes HOXA9 and MEIS1 expression. Massonianoside B exhibits anti-leukemic activity. It is metabolically stable in both human and mouse liver microsomes.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jie Chen et al.
ACS chemical biology, 14(5), 873-881 (2019-04-06)
Protein methyltransferases (PMTs) are involved in numerous biological processes and have been studied as a promising target class in the field of oncology and other diseases. Disruptor of telomeric silencing 1-like (DOT1L), a histone H3 lysine 79 (H3K79) methyltransferase, plays

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service