Skip to Content
Merck
All Photos(1)

Key Documents

SML3548

Sigma-Aldrich

8RK64

≥98% (HPLC)

Synonym(s):

(S)-N-(5-(2-azidoacetyl)-4,5,6,7-tetrahydrothiazolo[5,4-c]pyridin-2-yl)-1-cyanopyrrolidine-3-carboxamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$159.00
25 MG
$640.00

$159.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$159.00
25 MG
$640.00

About This Item

Empirical Formula (Hill Notation):
C14H16N8O2S
CAS Number:
Molecular Weight:
360.39
UNSPSC Code:
41121800
UNSPSC Code:
12352200
NACRES:
NA.21

$159.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C([C@H]1CCN(C#N)C1)NC2=NC3=C(S2)CN(C(CN=[N+]=[N-])=O)CC3

InChI key

KIWKRCCIHSGWQS-VIFPVBQESA-N

Biochem/physiol Actions

8RK64 is a cell penetrant, potent and selective covalent inhibitor of UCHL1 (ubiquitin carboxy-terminal hydrolase L1) that binds to the active-site cysteine residue of UCHL1. 8RK64 potently inhibits UCHL1 activity in cells.
Cell penetrant, potent and selective covalent inhibitor of UCHL1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Raymond Kooij et al.
Journal of the American Chemical Society, 142(39), 16825-16841 (2020-09-05)
Many reagents have emerged to study the function of specific enzymes in vitro. On the other hand, target specific reagents are scarce or need improvement, allowing investigations of the function of individual enzymes in their native cellular context. Here we

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service