Skip to Content
Merck
All Photos(1)

Key Documents

SML3527

Sigma-Aldrich

Ro 08-2750

≥95% (HPLC)

Synonym(s):

2,3,4,10-Tetrahydro-7,10-dimethyl-2,4-dioxobenzo[g]pteridine-8-carboxaldehyde, 7,10-Dimethyl-2,4-dioxo-2,3,4,10-tetrahydrobenzo[g]pteridine-8-carbaldehyde, 8a-Oxolumiflavin, Ro 082750, Ro08-2750

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$129.00
25 MG
$523.00

$129.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$129.00
25 MG
$523.00

About This Item

Empirical Formula (Hill Notation):
C13H10N4O3
CAS Number:
Molecular Weight:
270.24
MDL number:
UNSPSC Code:
51111800
UNSPSC Code:
12352200
NACRES:
NA.77

$129.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥95% (HPLC)

form

powder

color

, Light yellow to orange

solubility

DMSO: 2 mg/mL, clear (Warmed)

storage temp.

2-8°C

SMILES string

CC1=CC(N=C2C(N3)=O)=C(C=C1C=O)N(C)C2=NC3=O

InChI

1S/C13H10N4O3/c1-6-3-8-9(4-7(6)5-18)17(2)11-10(14-8)12(19)16-13(20)15-11/h3-5H,1-2H3,(H,16,19,20)

InChI key

JDEMVNYMYPJJIM-UHFFFAOYSA-N

Biochem/physiol Actions

Nerve growth factor (NGF) TrkA-/p75NTR (CD271)-binding and Musashi (MSI1, MSI2) RNA-binding antagonist (blocker, neutralizer).

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Muta. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service