Skip to Content
Merck
All Photos(1)

Key Documents

SML3514

Sigma-Aldrich

Corylin

≥98% (HPLC)

Synonym(s):

3-(2,2-Dimethyl-2H-1-benzopyran-6-yl)-7-hydroxy-4H-1-benzopyran-4-one

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$103.00
25 MG
$417.00

$103.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$103.00
25 MG
$417.00

About This Item

Empirical Formula (Hill Notation):
C20H16O4
CAS Number:
Molecular Weight:
320.34
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$103.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C1C2=CC=C(C=C2OC=C1C3=CC4=C(C=C3)OC(C)(C)C=C4)O

InChI key

PWAACAMQKVIVPZ-UHFFFAOYSA-N

Biochem/physiol Actions

Corylin is a flavonoid isolated from fruit and seed of medical herb Psoralea corylifolia that extend the replicative lifespan in yeast by targeting the Gtr1 protein. Also, Corylin extends lifespan in mice fed a high-fat diet. Corylin exhibits numerous pharmacological effects such as antioxidant properties, anti-inflammatory effects, osteoblastic proliferation-stimulating activity, insulin resistance amelioration.
Extend the replicative lifespan in yeast by targeting the Gtr1 protein

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

I-Chen Chen et al.
Pharmaceuticals (Basel, Switzerland), 14(10) (2021-10-24)
Acute lung injury (ALI) is a high mortality disease with acute inflammation. Corylin is a compound isolated from the whole plant of Psoralea corylifolia L. and has been reported to have anti-inflammatory activities. Herein, we investigated the therapeutic potential of
Zi-Yu Chang et al.
International journal of molecular sciences, 23(5) (2022-03-11)
Inflammatory bowel disease (IBD) involves chronic inflammation, loss of epithelial integrity, and gastrointestinal microbiota dysbiosis, resulting in the development of a colon cancer known as colitis-associated colorectal cancer (CAC). In this study, we evaluated the effects of corylin in a
Tong-Hong Wang et al.
Nature communications, 13(1), 1238-1238 (2022-03-11)
In the long history of traditional Chinese medicine, single herbs and complex formulas have been suggested to increase lifespan. However, the identification of single molecules responsible for lifespan extension has been challenging. Here, we collected a list of traditional Chinese

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service